3-Tolualdehyde ≥98%

Supplier: ACROS ORGANICS MS

Synonyms: m-Tolylaldehydem-Tolualdehyde3-Methylbenzaldehyde

AC13899-0250 AC13899-1000
200019-854EA 68.58 USD
200019-854 200013-416
3-Tolualdehyde ≥98%
3-Tolualdehyde
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 199 °C (1013 hPa)
Density: 1.0535 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003374
CAS Number: 620-23-5
EINECS: 210-632-0

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5420 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.1 % hydroquinone

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR