Thioacetic acid 98%

Danger

AC13878-5000 AC13878-0025
103525-522EA 711.92 USD
103525-522 200014-806
Thioacetic acid 98%
Thioacetic acid
Formula: C₂H₄OS
MW: 76.12 g/mol
Boiling Pt: 87…93 °C (760 mmHg)
Melting Pt: –17 °C
Density: 1.07 g/cm³
Flash Pt: 18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004853
CAS Number: 507-09-5
EINECS: 208-063-8
UN: 2436
ADR: 3,II
Merck Index: 12,09454

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR