1,3-Diphenyl-2-thiourea 98%

Warning

Synonyms: N,N’-DiphenylthioureaThiocarbanilideN,N'-Diphenylthiourea

AC13870-1000 AC13870-0025 AC13870-0010 AC13870-5000
200032-958EA 18.3 USD
200032-958 200014-802 200050-984 103530-982
1,3-Diphenyl-2-thiourea 98%
1,3-Diphenyl-2-thiourea
Formula: C₁₃H₁₂N₂S
MW: 228.32 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00004921
CAS Number: 102-08-9
EINECS: 203-004-2
UN: 3077
ADR: 9,III
Merck Index: 13,03366

Order Now

Specification Test Results

Appearance White to cream to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 148 to 154 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR