1-(2-Thenoyl)-3,3,3-trifluoroacetone 99%

Warning

Synonyms: Thenoyltrifluoroacetone2-Thenoyltrifluoroacetone4,4,4-Trifluoro-1-(2-thienyl)butane-1,3-dione(2-Thenoyl)-ω,ω,ω-trifluoroacetone(TTA)TTFA

AC13861-0250 AC13861-1000 AC13861-5000
200007-852EA 163 USD
200007-852 200020-920 103510-370
1-(2-Thenoyl)-3,3,3-trifluoroacetone 99%
1-(2-Thenoyl)-3,3,3-trifluoroacetone
Formula: C₈H₅F₃O₂S
MW: 222.19 g/mol
Boiling Pt: 96…98 °C (8 mmHg)
Melting Pt: 40…44 °C
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00005445
CAS Number: 326-91-0
EINECS: 206-316-7

Order Now

Specification Test Results

Appearance Light yellow to beige to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 40 to 44 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR