Tetramethylurea 99%

Danger

Synonyms: N,N,N',N'-Tetramethylurea

AC13849-1000 AC13849-0010 AC13849-2500
103510-364EA 504 USD
103510-364 200015-462 200027-528
Tetramethylurea 99%
Tetramethylurea
Formula: C₅H₁₂N₂O
MW: 116.16 g/mol
Boiling Pt: 174…178 °C (760 mmHg)
Melting Pt: –1 °C
Density: 0.9879 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00008319
CAS Number: 632-22-4
EINECS: 211-173-9
Merck Index: 12,09368

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4520 (20 °C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR