Sulfolane 99+%

Warning

Synonyms: Tetramethylene sulphoneTetramethylene sulfoneTetrahydrothiophene-1,1-dioxideTetrahydrothiophene 1,1-dioxide

AC13843-0100 AC13843-0025 AC13843-2500 AC13843-0010
103525-510EA 798 USD
103525-510 103525-508 200000-904 103525-506
Sulfolane 99+%
Sulpholane
Formula: C₄H₈O₂S
MW: 120.17 g/mol
Boiling Pt: 285 °C (760 mmHg)
Melting Pt: 27.4…27.8 °C
Density: 1.26 g/cm³
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00005484
CAS Number: 126-33-0
EINECS: 204-783-1
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4820 to 1.4850 (20 °C, 589 nm)
Water ≤0.3 %
Sulfated ash ≤0.1 %
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR