3,3,5,5-Tetramethylcyclohexanone 98%

AC13839-0500 AC13839-0100
103465-022EA 340 USD
103465-022
3,3,5,5-Tetramethylcyclohexanone 98%
3,3,5,5-Tetramethylcyclohexanone
Formula: C₁₀H₁₈O
Boiling Pt: 200…203 °C (760 mmHg)
Melting Pt: 11…12 °C
Density: 0.8954 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00001640
CAS Number: 14376-79-5
EINECS: 238-350-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20 °C, 589 nm)
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR