Tetrabutyltin 96%

Danger

Synonyms: TetrabutylstannaneTetra-n-butyltin

AC13798-1000 AC13798-5000
103530-970EA 93.3 USD
103530-970 200032-942
Tetrabutyltin 96%
Tetrabutyltin
Formula: C₁₆H₃₆Sn
MW: 347.17 g/mol
Boiling Pt: 145 °C (10 mmHg)
Melting Pt: –97 °C
Density: 1.057 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00009422
CAS Number: 1461-25-2
EINECS: 215-960-8
UN: 2788
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to straw yellow Liquid
Infrared spectrum Conforms
GC ≥95 %
Titration with NaOH 98.5 to 100.5 % (On anhydrous substance)
Color of solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR