Terephthalaldehyde 98%

Warning

Synonyms: Terephthaldicarboxaldehydep-PhthalaldehydeTerephthaldialdehyde

AC13789-1000 AC13789-5000
200044-716EA 216 USD
200044-716 200007-778
Terephthalaldehyde 98%
Terephthalaldehyde
Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 114…116 °C
Flash Pt: 76 °C
MDL Number: MFCD00006949
CAS Number: 623-27-8
EINECS: 210-784-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 114 to 116 °C
GC ≥97.5 %
Specific optical rotation +35° to +39° (20 °C, 589 nm) (C=5, 1N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR