3-Bromo-4-chlorobenzotrifluoride 98+%

Danger

Synonyms: 3-Bromo-4-chloro-α,α,αa-trifluorotoluene2-Bromo-1-chloro-4-(trifluoromethyl)benzene1-Bromo-2-chloro-5-(trifluoromethyl)benzene

AC13751-0250 AC13751-0050
200047-412EA 20.3 USD
200047-412 200047-410
3-Bromo-4-chlorobenzotrifluoride 98+%
3-Bromo-4-chlorobenzotrifluoride
Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
Boiling Pt: 188 °C (1013 hPa)
Melting Pt: 23…22 °C
Density: 1.74 g/cm³
Flash Pt: 94 °C
MDL Number: MFCD00018093
CAS Number: 454-78-4
EINECS: 207-226-0
UN: 1760

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4980 to 1.5000 (20 °C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR