3-Iodopyridine 99%
|
Danger
|
AC13744-0010
AC13744-0100
200017-114EA
30.5
USD
200017-114
200029-332
3-Iodopyridine 99%
3-Iodopyridine
Formula:
C₅H₄IN MW: 205 g/mol Boiling Pt: 90…92 °C (14 mmHg) Melting Pt: 53…56 °C Flash Pt: 106 °C |
MDL Number:
MFCD00023553 CAS Number: 1120-90-7 EINECS: 214-322-6 |
Specification Test Results
Appearance | Light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 53 to 56 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...