1-(2,4-Xylyl)piperazin 99%

Danger

Synonyms: 1-(2,4-Dimethylphenyl)piperazine1-(2,4-Xylyl)piperazine

AC13694-0050
200008-302EA 41.3 USD
200008-302
1-(2,4-Xylyl)piperazin 99%
1-(2,4-Xylyl)piperazin
Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
Boiling Pt: 153 °C (10 mmHg)
MDL Number: MFCD00023127
CAS Number: 1013-76-9
EINECS: 213-797-7
UN: 2735

Order Now

Specification Test Results

Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.5530 to 1.5550 (20 °C, 589 nm)
GC ≥98.5 %
Titration with HClO4 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR