2-Methyl-2-nitropropane-1,3-diol 99%

Synonyms: 2-Methyl-2-nitro-1,3-propanediol2-Nitro-2-methyl-1,3-propanediol

AC13480-0050
200034-938EA 78.8 USD
200034-938
2-Methyl-2-nitropropane-1,3-diol 99%
2-Methyl-2-nitropropane-1,3-diol
Formula: C₄H₉NO₄
MW: 135.12 g/mol
Boiling Pt: 324 °C (1013 hPa)
Melting Pt: 147…149 °C
MDL Number: MFCD00024812
CAS Number: 77-49-6
EINECS: 201-031-4

Order Now

Specification Test Results

Appearance White to yellow Crystals
Infrared spectrum Conforms
GC ≥98.5 %
NMR (31P-NMR) ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR