1-Cyclohexylpiperazine 98%

Danger

AC13430-0050 AC13430-0250
200004-720EA 39.4 USD
200004-720 200021-810
1-Cyclohexylpiperazine 98%
1-Cyclohexylpiperazine
Formula: C₁₀H₂₀N₂
MW: 168.28 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 28…32 °C
MDL Number: MFCD00044809
CAS Number: 17766-28-8
EINECS: 241-750-0
UN: 3259

Order Now

Specification Test Results

Appearance Yellow-brown to brown Low melting solid
Infrared spectrum Conforms
GC ≥97.5 %
GC ≥99.0 %
Residue after evaporation ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR