Suberic acid 99%

Warning

Synonyms: Octanedioic acid1,6-Hexanedicarboxylic acid1,8-Octanedioic acid

AC13281-5000 AC13281-1000 AC13281-0025
200020-746EA 126 USD
200020-746 200020-748 103510-308
Suberic acid 99%
Suberic acid
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 230 °C (15 mmHg)
Melting Pt: 141…144 °C
Flash Pt: 203 °C
Storage Temperature: Ambient
MDL Number: MFCD00004428
CAS Number: 505-48-6
EINECS: 208-010-9
Merck Index: 12,09031

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 141 to 144 °C
Titration with NaOH 98.5 to 101.5 %
Elemental Analysis 6.22 to 7.02 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR