Spermidine 99%

Danger

Synonyms: N1-(3-Aminopropyl)butane-1,4-diamineN-(3-Aminopropyl)-1,4-diaminobutane4-Azaoctamethylenediamine

AC13274-0250 AC13274-0010 AC13274-1000 AC13274-0050
103525-458EA 524 USD
103525-458 200000-862 103525-460 200007-732
Spermidine 99%
Spermidine
Formula: C₇H₁₉N₃
MW: 145.25 g/mol
Boiling Pt: 128…130 °C (14 mmHg)
Melting Pt: 22…25 °C
Density: 0.925 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008229
CAS Number: 124-20-9
EINECS: 204-689-0
UN: 3259
ADR: 8,II
Merck Index: 12,08893

Order Now

Specification Test Results

Appearance Clear colorless Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4785 to 1.4805 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR