Pyruvic acid 98%, Extra Pure

Danger

Synonyms: 2-Oxopropionic acid2-Oxopropanoic acid2-Ketopropionic acidα-Ketopropionic acid

AC13214-0250 AC13214-5000 AC13214-0025 AC13214-1000
103525-444EA 452 USD
103525-444 200000-834 200000-838 200000-836
Pyruvic acid 98%, Extra Pure
Pyruvic acid
Formula: C₃H₄O₃
MW: 88.06 g/mol
Boiling Pt: 165 °C (760 mmHg)
Melting Pt: 11.8 °C
Density: 1.25 g/cm³
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002585
CAS Number: 127-17-3
EINECS: 204-824-3
UN: 3265
ADR: 8,II
Merck Index: 12,08205

Order Now

Specification Test Results

Appearance Clear colorless to yellow to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4320 (20 °C, 589 nm)
Specific gravity (20 °C) 1.240 to 1.280
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR