Pyridine-3,5-dicarboxylic acid 98%

Warning

Synonyms: 3,5-Pyridinedicarboxylic acidDinicotinic acid

AC13190-0100 AC13190-0500
103530-934EA 341 USD
103530-934 200016-910
Pyridine-3,5-dicarboxylic acid 98%
Pyridine-3,5-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006393
CAS Number: 499-81-0
EINECS: 207-893-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR