Propylene carbonate 99.5%

Warning

Synonyms: 4-Methyl-1,3-dioxolan-2-one

AC13156-0010 AC13156-0025 AC13156-5000
200007-510EA 264.23 USD
200007-510 200007-512 103467-236
Propylene carbonate 99.5%
Propylene carbonate
Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 240…243 °C (760 mmHg)
Melting Pt: –49 °C
Density: 1.204 g/cm³
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00005385
CAS Number: 108-32-7
EINECS: 203-572-1

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4220 (20 °C, 589 nm)
Water ≤0.2 %
GC ≥99.4 %
Color scale ≤40 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR