Piperonyl alcohol 98%

Synonyms: 3,4-(Methylenedioxy)benzyl alcohol1,3-Benzodioxol-5-ylmethanol

AC13135-0250 AC13135-1000
200026-484EA 99.2 USD
200026-484 103510-256
Piperonyl alcohol 98%
Piperonyl alcohol
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 156…158 °C (66 mmHg)
Melting Pt: 51…55 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005836
CAS Number: 495-76-1
EINECS: 207-808-4

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 51 to 55 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR