(1S)-(-)-α-Pinene 98%

Danger

Synonyms: (-)-α-Pinene(1S)-(-)-2-PineneL(-)-α-Pinene(-)-Pin-2(3)-ene

AC13127-0025 AC13127-2500 AC13127-0050 AC13127-5000
103530-908EA 293 USD
103530-908 200015-198 200021-200 200021-202
(1S)-(-)-α-Pinene 98%
(1S)-(-)-α-Pinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 155…156 °C (760 mmHg)
Melting Pt: –64 °C
Density: 0.8595 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00064145
CAS Number: 7785-26-4
EINECS: 232-077-3
UN: 2368
ADR: 3,III
Merck Index: 12,07599

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4640 to 1.4680 (20 °C, 589 nm)
GC ≥97.5 %
Color scale ≤30 APHA
Specific gravity (20 °C) 0.858 to 0.861
Specific optical rotation -35° to -45° (20 °C, 589 nm) (Neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR