(1R)-(+)-α-Pinene 98%, 80% ee

Danger

Synonyms: (1R)-(+)-2-Pinene(+)-Pin-2(3)-eneD(+)-α-Pinene(+)-α-Pinene

AC13126-1000 AC13126-5000 AC13126-2500
200015-212EA 64.1 USD
200015-212 200015-210 200021-216
(1R)-(+)-α-Pinene 98%, 80% ee
(1R)-(+)-α-Pinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.86 g/cm³
Flash Pt: 33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001346
CAS Number: 7785-70-8
EINECS: 232-087-8
UN: 2368
ADR: 3,III
Merck Index: 12,07599

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4650 to 1.4670 (20 °C, 589 nm)
GC ≥97.5 %
Enantiomeric excess ≥75 %
Specific gravity (20 °C) 0.855 to 0.865
Specific optical rotation +40° to +47° (20 °C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR