Pimelic acid 98%

Warning

Synonyms: 1,5-Pentanedicarboxylic acidHeptanedioic acid1,7-Heptanedioic acid

AC13123-5000 AC13123-0250 AC13123-1000
200014-060EA 65.8 USD
200014-060 103466-318 200007-364
Pimelic acid 98%
Pimelic acid
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 212 °C (10 mmHg)
Melting Pt: 103…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00004425
CAS Number: 111-16-0
EINECS: 203-840-8
Merck Index: 12,07585

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 103 to 106 °C
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR