Picolinic acid 99%

Warning

Synonyms: 2-Pyridinecarboxylic acid2-Picolinic acidPyridine-2-carboxylic acid

AC13117-0250 AC13117-5000 AC13117-1000
200050-480EA 23 USD
200050-480 200007-362 200020-404
Picolinic acid 99%
Picolinic acid
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Melting Pt: 136…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00006293
CAS Number: 98-98-6
EINECS: 202-719-7
Merck Index: 12,07557

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 139 °C
Titration with NaOH 98.5 to 101.5 %
Residue after evaporation ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR