3-Picoline-N-oxide 98%

Warning

Synonyms: 3-Picoline N-oxide3-Methylpyridine 1-oxide3-Methylpyridine-1-oxide3-Methylpyridine-N-oxide3-Picoline 1-oxide

AC13115-1000 AC13115-5000
200017-200EA 40.37 USD
200017-200 200029-436
3-Picoline-N-oxide 98%
3-Picoline N-oxide
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 150 °C (15 mmHg)
Melting Pt: 38…40 °C
Flash Pt: 143 °C
MDL Number: MFCD00006203
CAS Number: 1003-73-2
EINECS: 213-714-4

Order Now

Specification Test Results

Appearance Light yellow to brown Low melting crystalline mass
Infrared spectrum Conforms
GC ≥97.5 %
GC ≥99.75 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR