4-Phenylpyridine 99%

Warning

AC13091-0250 AC13091-0050
200023-984EA 254 USD
200023-984 200023-982
4-Phenylpyridine 99%
4-Phenylpyridine
Formula: C₁₁H₉N
MW: 155.2 g/mol
Boiling Pt: 274…275 °C (760 mmHg)
Melting Pt: 71…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00006420
CAS Number: 939-23-1
EINECS: 213-357-4

Order Now

Specification Test Results

Appearance White to light yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 71 to 75 °C
GC ≥98.5 %
Residue after evaporation ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR