2-Phenylindole 99%

Synonyms: 2-Phenyl-1H-indole

AC13068-0050 AC13068-1000 AC13068-5000
200047-082EA 29.2 USD
200047-082 200022-906 200034-982
2-Phenylindole 99%
2-Phenylindole
Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Boiling Pt: 250 °C (10 mmHg)
Melting Pt: 188…191 °C
MDL Number: MFCD00005608
CAS Number: 948-65-2
EINECS: 213-436-3

Order Now

Specification Test Results

Appearance Off-white to beige to light green Powder
Infrared spectrum Conforms
Melting point 188 to 191 °C
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.2 %
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR