N-Phenylethylenediamine 99%

Danger

AC13061-0100 AC13061-0500
103530-896EA 208 USD
103530-896 200013-746
N-Phenylethylenediamine 99%
N-Phenylethylenediamine
Formula: C₆H₅NHCH₂CH₂NH₂
MW: 136.2 g/mol
Boiling Pt: 262…264 °C (760 mmHg)
Density: 1.04 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00008162
CAS Number: 1664-40-0
EINECS: 216-773-4
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5855 to 1.5875 (20 °C, 589 nm)
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR