Diphenyl ether 99%

Warning

Synonyms: Diphenyl oxidePhenyl etherPhenoxybenzene

AC13060-0100 AC13060-5000 AC13060-0025 AC13060-1000 AC13060-0010
103526-014EA 269.57 USD
103526-014 200000-776 200000-778 103525-374 103525-370
Diphenyl ether 99%
Diphenyl ether
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 259 °C (760 mmHg)
Melting Pt: 26…30 °C
Density: 1.073 g/cm³
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00003034
CAS Number: 101-84-8
EINECS: 202-981-2
UN: 3077
ADR: 9,III
Merck Index: 12,07442

Order Now

Specification Test Results

Appearance Clear light yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5795 to 1.5815 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR