Phenothiazine 99%

Warning

Synonyms: 10H-Phenothiazine

AC13021-0050 AC13021-5000 AC13021-0010
200020-352EA 78.7 USD
200020-352 103466-292 200020-354
Phenothiazine 99%
Phenothiazine
Formula: C₁₂H₉NS
MW: 199.28 g/mol
Boiling Pt: 371 °C
Melting Pt: 185 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00005015
CAS Number: 92-84-2
EINECS: 202-196-5
UN: 3077
ADR: 9,III
Merck Index: 12,07404

Order Now

Specification Test Results

Appearance Yellow Beads or prills
Infrared spectrum Conforms
Melting point ≥184 °C
HPLC ≥98.5 %
UV at 210 nm A: ≤0.05
UV at 220 nm A: ≤0.03
UV at 230 nm A: ≤0.02
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.2 M soln.) Passes test
Suitability (HPLC gradient) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR