Phenazine 98%

Warning

AC13015-0100 AC13015-1000 AC13015-0050
200032-504EA 42.11 USD
200032-504 200020-348 103466-280
Phenazine 98%
Phenazine
Formula: C₁₂H₈N₂
MW: 180.21 g/mol
Boiling Pt: >360 °C (760 mmHg)
Melting Pt: 174…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00005023
CAS Number: 92-82-0
EINECS: 202-193-9
Merck Index: 12,07359

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 173 to 177 °C
HPLC ≥97.5 %
UV at 210 nm A: ≤0.1
UV at 220 nm A: ≤0.06
UV at 230 nm A: ≤0.04
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR