Phenanthrene 97%

Warning

AC13009-0500 AC13009-0050 AC13009-5000
200007-306EA 382 USD
200007-306 200007-304 200013-956
Phenanthrene 97%
Phenanthrene
Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 95…101 °C
Density: 1.063 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00001168
CAS Number: 85-01-8
EINECS: 201-581-5
UN: 3077
ADR: 9,III
Merck Index: 12,07354
https://us.vwr.com/stibo/web/std.lang.all/84/56/2298456.jpg

Order Now

Specification Test Results

Appearance White to grey to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 97 to 101 °C
GC ≥96.0 %
Stabilizer 150 to 400 ppm BHT
Peroxides ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR