Perylene 99+%

AC13007-0010 AC13007-0050
103509-482EA 130 USD
103509-482 200013-944
Perylene 99+%
Perylene
Formula: C₂₀H₁₂
MW: 252.32 g/mol
Melting Pt: 277…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00004142
CAS Number: 198-55-0
EINECS: 205-900-9
Merck Index: 12,07325

Order Now

Specification Test Results

Appearance Ochre to brown Powder
Infrared spectrum Conforms
Melting point 277 to 280 °C
HPLC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR