Acetylacetone 99+%

Danger

Synonyms: 2,4-PentanedioneACAC

AC12996-0050
103529-076EA 33.9 USD
103529-076
Acetylacetone 99+%
Acetylacetone
Formula: CH₃COCH₂COCH₃
MW: 100.12 g/mol
Boiling Pt: 133…135 °C (760 mmHg)
Melting Pt: –23 °C
Density: 0.975 g/cm³
Flash Pt: 35.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008787
CAS Number: 123-54-6
EINECS: 204-634-0
UN: 2310
ADR: 3,III
Merck Index: 13,00083
https://us.vwr.com/stibo/web/std.lang.all/15/18/721518.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20 °C, 589 nm)
Water ≤0.1 %
GC ≥99.0 %
Color scale ≤50 APHA
Specific gravity (20 °C) 0.972 to 0.978
Acidity (CH3COOH) ≤0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR