Pentafluorothiophenol 97%

Danger

Synonyms: Pentafluorophenyl mercaptan2,3,4,5,6-PentafluorothiophenolPentafluorobenzenethiolMercaptopentafluorobenzene

AC12992-0250 AC12992-0010 AC12992-0050
103525-342EA 377 USD
103525-342 200020-326 103525-340
Pentafluorothiophenol 97%
Pentafluorothiophenol
Formula: C₆HF₅S
MW: 200.13 g/mol
Boiling Pt: 143 °C (760 mmHg)
Melting Pt: –24 °C
Density: 1.5 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00004828
CAS Number: 771-62-0
EINECS: 212-236-3
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4630 to 1.4650 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR