1,4-Pentadiene 99%

Danger

Synonyms: Penta-1,4-diene

AC12986-0010 AC12986-0050
103530-878EA 35.4 USD
103530-878 200008-516
1,4-Pentadiene 99%
1,4-Pentadiene
Formula: C₅H₈
MW: 68.12 g/mol
Boiling Pt: 26 °C (760 mmHg)
Melting Pt: –148 °C
Density: 0.65 g/cm³
Flash Pt: –20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008665
CAS Number: 591-93-5
EINECS: 209-736-9
UN: 3295
ADR: 3,I

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3884 to 1.3904 (20 °C, 589 nm)
GC ≥98.5 %
Specific optical rotation +63° to +67° (20 °C, 589 nm) (c=4, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR