Oxalic acid dihydrate 99+%, Extra Pure

Danger

Synonyms: Ethanedioic acid dihydrate

AC12960-0010 AC12960-1000 AC12960-0050
103525-336EA 33.3 USD
103525-336 200000-740 103525-334
Oxalic acid dihydrate 99+%, Extra Pure
Oxalic acid dihydrate
Formula: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
ADR: 8,III
REACH: 01-2119534576-33
Merck Index: 12,07043
https://us.vwr.com/stibo/web/std.lang.all/72/16/2297216.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 102 °C
Sulfated ash ≤0.1 %
Titration with KMnO4 ≥99.0 %
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR