Octanoyl chloride 99%

Danger

AC12943-1000 AC12943-0025 AC12943-5000
103530-876EA 397.38 USD
103530-876 200013-782 200013-784
Octanoyl chloride 99%
Octanoyl chloride
Formula: C₈H₁₅ClO
MW: 162.66 g/mol
Boiling Pt: 195 °C (760 mmHg)
Melting Pt: –63 °C
Density: 0.95 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00000766
CAS Number: 111-64-8
EINECS: 203-891-6
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20 °C, 589 nm)
Titration Argentometric ≥98.5 %
GC ≥98.5 %
Color scale ≤80 APHA
Acidity ≤1 %
Phosphorus (P) ≤150 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR