2-Octanone 99+%

Warning

Synonyms: Octan-2-one

AC12942-0250 AC12942-0025 AC12942-5000
200005-136EA 74.5 USD
200005-136 103525-328 200028-922
2-Octanone 99+%
2-Octanone
Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 173 °C (760 mmHg)
Melting Pt: –16 °C
Density: 0.819 g/cm³
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00009540
CAS Number: 111-13-7
EINECS: 203-837-1
UN: 1224
ADR: 3,III
Merck Index: 13,04731

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4160 (20 °C, 589 nm)
GC ≥99.0 %
Clarity of solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR