Norharmane 98%

Synonyms: Norharman9H-Pyrido[3,4-b]indole

AC12926-5000 AC12926-0025
103510-180EA 399 USD
103510-180 200007-200
Norharmane 98%
Norharmane
Formula: C₁₁H₈N₂
MW: 168.2 g/mol
Melting Pt: 199…201 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004956
CAS Number: 244-63-3
EINECS: 205-959-0

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 198 to 202 °C
Titration Tertiary base ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR