n-Nonane 99%, pure

Danger

AC12911-0025 AC12911-5000 AC12911-1000
200000-720EA 478.85 USD
200000-720 103524-082 200000-718
n-Nonane 99%, pure
n-Nonane
Formula: H₃C(CH₂)₇CH₃
MW: 128.26 g/mol
Boiling Pt: 151 °C (760 mmHg)
Melting Pt: –53 °C
Density: 0.718 g/cm³
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00009574
CAS Number: 111-84-2
EINECS: 203-913-4
UN: 1920
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4060 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR