4-Nitrotoluene 99%

Danger

AC12905-0010 AC12905-0050 AC12905-1000
200036-056EA 27 USD
200036-056 200029-952 200005-526
4-Nitrotoluene 99%
4-Nitrotoluene
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 238.3 °C (1013 hPa)
Melting Pt: 51…54 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00007366
CAS Number: 99-99-0
EINECS: 202-808-0
UN: 3446
ADR: 6.1,II
Merck Index: 12,06748

Order Now

Specification Test Results

Appearance Yellow Crystalline solid
Infrared spectrum Conforms
Melting point 51 to 54 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR