5-Nitroisatin 97%

Warning

Synonyms: 5-Nitro-1H-indole-2,3-dione5-Nitroindoline-2,3-dione

AC12872-0250 AC12872-0050
200048-252EA 102 USD
200048-252 200030-140
5-Nitroisatin 97%
5-Nitroisatin
Formula: C₈H₄N₂O₄
MW: 192.13 g/mol
Melting Pt: 251 °C (dec.)
MDL Number: MFCD00005720
CAS Number: 611-09-6
EINECS: 210-252-5

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
GC ≥98.5 %
Stabilizer Approx. 500 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR