2-Nitrobenzaldehyde 99+%

Warning

AC12840-5000 AC12840-1000 AC12840-0250
103463-582EA 786 USD
103463-582 200005-130 200005-132
2-Nitrobenzaldehyde 99+%
2-Nitrobenzaldehyde
Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 153 °C (23 mmHg)
Melting Pt: 41…44 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007132
CAS Number: 552-89-6
EINECS: 209-025-3
Merck Index: 12,06684

Order Now

Specification Test Results

Appearance Yellow Powder or needles and/or chunks
Infrared spectrum Conforms
Melting point 41 to 45 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR