4-Nitroaniline 99%

Danger

Synonyms: pNAp-Nitroaniline

AC12837-1000 AC12837-5000 AC12837-0025
200048-012EA 72.26 USD
200048-012 103524-076 103524-074
4-Nitroaniline 99%
4-Nitroaniline
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Boiling Pt: 332 °C (760 mmHg)
Melting Pt: 146…151 °C
Flash Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00007858
CAS Number: 100-01-6
EINECS: 202-810-1
UN: 1661
ADR: 6.1,II
Merck Index: 12,06681

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 147 to 151 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR