1-Naphthol 99+%

Danger

Synonyms: α-Naphtholα-Hydroxynaphthalene1-Hydroxynaphthalene

AC12819-5000 AC12819-0025 AC12819-1000
200000-708EA 69.9 USD
200000-708 103524-072 200008-740
1-Naphthol 99+%
1-Naphthol
Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 278…280 °C (760 mmHg)
Melting Pt: 95…97 °C
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00003930
CAS Number: 90-15-3
EINECS: 201-969-4
UN: 2811
ADR: 6.1,III
Merck Index: 12,06470

Order Now

Specification Test Results

Appearance Beige to grey to brown Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 95 to 97 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR