2-Naphthaldehyde 98%

Warning

Synonyms: β-Naphthaldehyde

AC12810-1000 AC12810-0250 AC12810-0050
103524-070EA 350 USD
103524-070 200063-368 200009-540
2-Naphthaldehyde 98%
2-Naphthaldehyde
Formula: C₁₁H₈O
MW: 156.18 g/mol
Boiling Pt: 160 °C (19 mmHg)
Melting Pt: 58…62 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004094
CAS Number: 66-99-9
EINECS: 200-640-2
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance Off-white to yellow to light beige Powder and/or chunks
Infrared spectrum Conforms
Melting point 58 to 62 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR