Methylurea 97%

Synonyms: MethylcarbamideN-Methylurea1-Methylurea

AC12799-5000 AC12799-1000
200025-928EA 92.2 USD
200025-928 200019-806
Methylurea 97%
Methylurea
Formula: C₂H₆N₂O
MW: 74.08 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00007950
CAS Number: 598-50-5
EINECS: 209-935-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
Water ≤2 %
HPLC ≥96.0 % (on dry substance)
Titration with HClO4 ≥96.0 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR