2-Methylthiophene 98%

Danger

AC12785-0250 AC12785-1000
200009-538EA 45.5 USD
200009-538 103530-840
2-Methylthiophene 98%
2-Methylthiophene
Formula: C₅H₆S
MW: 98.17 g/mol
Boiling Pt: 113 °C (760 mmHg)
Melting Pt: –63 °C
Density: 1.02 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00005451
CAS Number: 554-14-3
EINECS: 209-063-0
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5210 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR