α-Methylstyrene 99% stabilized
|
Danger
|
Synonyms:
Isopropenylbenzene
, 2-Phenylpropene
AC12771-0010
AC12771-0025
AC12771-1000
200000-694EA
51.9
USD
200000-694
200005-688
103524-054
α-Methylstyrene 99% stabilized
α-Methylstyrene
Formula:
C₉H₁₀ MW: 118.18 g/mol Boiling Pt: 165…169 °C (760 mmHg) Melting Pt: –23 °C Density: 0.909 g/cm³ Flash Pt: 45 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008859 CAS Number: 98-83-9 EINECS: 202-705-0 UN: 2303 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5370 to 1.5390 (20 °C, 589 nm) |
GC | ≥98.5 % |
Stabilizer | 15 ppm tert.-butylcatechol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...